missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 97%, Thermo Scientific™

Product Code. 10357104
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10357104 250 mg Amber Glass Bottle 250mg
10542264 1 g Amber Glass Bottle 1g
11306795 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357104 Supplier Thermo Scientific Maybridge Supplier No. CC55639CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-39-5
Molecular Formula C17H26BNO2
Molecular Weight (g/mol) 287.21
MDL Number MFCD08690299
InChI Key AGSIDMRVRGPBIE-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl pyrrolidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine, 4-pyrrolidine methyl phenylboronic acid pinacol ester, pyrrolidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 4-pyrrolidinomethyl phenylboronic acid, pinacol ester, amtb121, 4-1-pyrroldinylmethyl benzeneboronic acid pinacol ester, 4-pyrrolidin-1-ylmethyl phenylboronic acid pinacol ester
PubChem CID 18525868
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]pyrrolidine
Formula Weight 287.21
Percent Purity 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.