missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4-hydroxyphenyl)piperazino]ethan-1-one, Thermo Scientific™

Product Code. 10795853
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10795853 50 g 50g
10557864 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10795853 Supplier Thermo Scientific Maybridge Supplier No. SB02047EE
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67914-60-7
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD00044905
InChI Key AGVNLFCRZULMKK-UHFFFAOYSA-N
Synonym 1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
PubChem CID 712441
IUPAC Name 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1

Specifications

Assay Percent Range 97%
Quantity 50 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.