missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(5-phenyloxazol-2-yl)benzene, 98+%, Thermo Scientific™

Product Code. 11439433
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439433 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11439433 Supplier Thermo Scientific Alfa Aesar Supplier No. A12321.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1806-34-4
Melting Point 241°C to 243°C
Molecular Formula C24H16N2O2
MDL Number MFCD00005309
Quantity 25 g
Beilstein 325741
Synonym popop, 1,4-bis 5-phenyloxazol-2-yl benzene, oxazole, 2,2'-1,4-phenylene bis 5-phenyl, p-bis 5-phenyloxazol-2-yl benzene, 2,2'-p-phenylenebis 5-phenyloxazole, 1,4-bis 5-phenyl-2-oxazolyl benzene, 5-phenyl-2-4-5-phenyl-1,3-oxazol-2-yl phenyl-1,3-oxazole, 1,4-di 5-phenyl-2-oxazolyl benzene, 1,4-bis 2-5-phenyloxazolyl benzene, oxazole, 2,2'-p-phenylenebis 5-phenyl
InChI Key MASVCBBIUQRUKL-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=C(C=C3)C4=NC=C(O4)C5=CC=CC=C5
IUPAC Name 5-phenyl-2-[4-(5-phenyl-1,3-oxazol-2-yl)phenyl]-1,3-oxazole
Molecular Weight (g/mol) 364.404
PubChem CID 15732
ChEBI CHEBI:52236
Formula Weight 364.41
Percent Purity ≥98%
Chemical Name or Material 1,4-Bis (5-phenyloxazol-2-yl)benzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.