missing translation for 'onlineSavingsMsg'
Learn More

1-(4-bromobutoxy)-4-nitrobenzene, 85%, Thermo Scientific™

Product Code. p-3211266 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10660925

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 55502-03-9
Molecular Formula C10H12BrNO3
Molecular Weight (g/mol) 274.11
MDL Number MFCD00980317
InChI Key DBRBCFJUEVSKGZ-UHFFFAOYSA-N
Synonym 1-4-bromobutoxy-4-nitrobenzene, 4-nitrophenoxybutyl bromide
PubChem CID 2063669
IUPAC Name 1-(4-bromobutoxy)-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OCCCCBr

Specifications

CAS Min % 80.0
CAS Max % 100.0
Melting Point 37°C to 40°C
Color Beige to Brown
Boiling Point 165°C to 170°C (0.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H4O(CH2)4Br
Quantity 5 g
Formula Weight 274.11
Percent Purity 85%
Physical Form Adhering Chunks
Chemical Name or Material 1-(4-Bromobutoxy)-4-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Bromobutoxy)-4-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.