missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-(4-Bromophenyl)ethylamine, 96%

Product Code. 11392657
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392657 1 g 1g
11302667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392657 Supplier Thermo Scientific Alfa Aesar Supplier No. L11848.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24358-62-1 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24358-62-1
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00025548
InChI Key SOZMSEPDYJGBEK-UHFFFAOYSA-N
Synonym 1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
PubChem CID 91175
IUPAC Name 1-(4-bromophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Br)N

Specifications

Density 1.397
Boiling Point 140°C to 145°C (30 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.567
Quantity 1 g
UN Number UN2735
Beilstein 2412318
Sensitivity Air Sensitive
Formula Weight 200.08
Percent Purity 96%
Chemical Name or Material (+/-)-1-(4-Bromophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.