missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-methylethylamine, 97%

Product Code. 15455257
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15455257

Brand: Thermo Scientific Alfa Aesar H32527.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17797-11-4 | C9H12ClN | 169.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17797-11-4
Molecular Formula C9H12ClN
Molecular Weight (g/mol) 169.65
MDL Number MFCD08669636
InChI Key JCUUNSQUTQELEZ-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl propan-2-amine, 1-4-chlorophenyl-1-methylethylamine, 2-4-chlorophenyl prop-2-ylamine, 4-chloro-alpha,alpha-dimethylbenzylamine, 4-chlorocumylamine, alpha-amino-4-chlorocumene, 2-4-chlorophenyl-2-propanamine, 2-?4-chlorophenyl?propan-2-amine, benzenemethanamine,4-chloro-, a,, a-dimethyl, benzenemethanamine,4-chloro-a,a-dimethyl
PubChem CID 11030256
IUPAC Name 2-(4-chlorophenyl)propan-2-amine
SMILES CC(C)(N)C1=CC=C(Cl)C=C1

Specifications

Boiling Point 43°C to 45°C (0.3 mmHg)
Odor Amine-like
Quantity 250 mg
UN Number UN2735
Formula Weight 169.66
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-1-methylethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.