missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-chlorophenyl)sulfonyl]-1H-pyrrole, 97%, Thermo Scientific™

Product Code. 10386434
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10386434 25 g 25g
10233062 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386434 Supplier Thermo Scientific Maybridge Supplier No. BTB07116EB
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16851-83-5
Molecular Formula C10H8ClNO2S
Molecular Weight (g/mol) 241.689
MDL Number MFCD00067753
InChI Key ACNNPPGRQRFTSO-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl sulfonyl-1h-pyrrole, 1-4-chlorobenzenesulfonyl pyrrole, maybridge1_001769, cbmicro_045913, 1-4-chlorophenyl sulfonylpyrrole, 1-chloro-4-pyrrolylsulfonyl benzene, 4-chloro-1-pyrrolylsulfonyl benzene, 1-4-chlorobenzenesulfonyl-1h-pyrrole, 1-4-chlorophenylsulfonyl-1h-pyrrole
PubChem CID 706366
IUPAC Name 1-(4-chlorophenyl)sulfonylpyrrole
SMILES C1=CN(C=C1)S(=O)(=O)C2=CC=C(C=C2)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 241.7
Percent Purity 97%
Chemical Name or Material 1-[(4-chlorophenyl)sulfonyl]-1H-pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.