missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-chlorophenyl)sulfonyl]acetone, Thermo Scientific™

Product Code. 10245864
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10245864 5 g Amber Glass Bottle 5g
10061794 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245864 Supplier Thermo Scientific Maybridge Supplier No. KM01166DE
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5000-48-6
Molecular Formula C9H9ClO3S
Molecular Weight (g/mol) 232.68
MDL Number MFCD00018663
InChI Key BRDBHPZILGTBFY-UHFFFAOYSA-N
Synonym 4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
PubChem CID 521269
IUPAC Name 1-(4-chlorophenyl)sulfonylpropan-2-one
SMILES CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 232.68
Percent Purity 97%
Chemical Name or Material 1-[(4-chlorophenyl)sulfonyl]acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.