missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobenzene, 99%

Product Code. 10366220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
2.5kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366220 1 kg Plastic bottle 1kg
10010750 5 g Glass Bottle 5g
10020750 250 g Plastic bottle 250g
10043373 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10366220 Supplier Thermo Scientific Supplier No. 112730010
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-37-6
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000089
InChI Key SWJPEBQEEAHIGZ-UHFFFAOYSA-N
Synonym p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
PubChem CID 7804
ChEBI CHEBI:37150
IUPAC Name 1,4-dibromobenzene
SMILES BrC1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 86°C to 89°C
Color Colorless to White
Density 1.8400g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H4Br2
Quantity 1 kg
Beilstein 05, 211
Merck Index 15, 3024
Formula Weight 235.91
Percent Purity ≥98.5%
Physical Form Crystals
Chemical Name or Material 1, 4-Dibromobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.