missing translation for 'onlineSavingsMsg'
Learn More

1,4-Difluorobenzene, 99+%

Product Code. 10665532 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10665532

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-36-3 | C6H4F2 | 114.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-36-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000344
InChI Key QUGUFLJIAFISSW-UHFFFAOYSA-N
Synonym p-difluorobenzene, benzene, 1,4-difluoro, para-difluorobenzene, benzene, p-difluoro, 2,5-difluorobenzene, unii-y9fq2ef0z6, 1,4-difluorbenzol, y9fq2ef0z6, benzene,4-difluoro, pubchem2309
PubChem CID 10892
ChEBI CHEBI:38585
IUPAC Name 1,4-difluorobenzene
SMILES FC1=CC=C(F)C=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -13°C
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 88°C to 89°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H4F2
Refractive Index 1.4400 to 1.442
Quantity 25 mL
Beilstein 05, 199
Merck Index 15, 3167
Specific Gravity 1.17
Formula Weight 114.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 4-Difluorobenzene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Difluorobenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.