missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodobenzene, 98%

Product Code. 11418246
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418246 25 g 25g
11428246 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418246 Supplier Thermo Scientific Alfa Aesar Supplier No. A15335.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 624-38-4 | C6H4I2 | 329.907 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-38-4
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.907
MDL Number MFCD00001054
InChI Key LFMWZTSOMGDDJU-UHFFFAOYSA-N
Synonym p-diiodobenzene, benzene, 1,4-diiodo, p-benzene diiodide, benzene, p-diiodo, unii-i9xcf83vav, i9xcf83vav, 1,4-diiodbenzene, 1,4-diidobenzene, 1,4 diiodobenzene, 1,4-diiodiobenzene
PubChem CID 12208
IUPAC Name 1,4-diiodobenzene
SMILES C1=CC(=CC=C1I)I

Specifications

Melting Point 128°C to 132°C
Boiling Point 285°C
Quantity 25 g
Beilstein 1904546
Sensitivity Light sensitive
Solubility Information Soluble in ether and ethanol. Insoluble in water.
Formula Weight 329.91
Percent Purity 98%
Chemical Name or Material 1,4-Diiodobenzene
compliance-icons
Product Identifier
  • 1,4-Diiodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Diiodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.