missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, 99+%

Product Code. 10182710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10182710 100 g Glass bottle 100g
10589240 500 g Glass bottle 500g
10409220 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10182710 Supplier Thermo Scientific Acros Supplier No. 115411000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 150-78-7 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-78-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008401
InChI Key OHBQPCCCRFSCAX-UHFFFAOYSA-N
Synonym p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
PubChem CID 9016
IUPAC Name 1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 54°C to 58°C
Color White
Density 1.0500g/mL
Boiling Point 213°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(OCH3)2
Quantity 100 g
Beilstein 06, 843
Specific Gravity 1.05
Solubility Information Solubility in water: 0.8g/L (20 c). Other solubilities: soluble in benzene,alcohol,ethylacetate and,acetone,1g/10mL dioxane
Viscosity 1.04 mPa.s (65°C)
Formula Weight 138.17
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 1, 4-Dimethoxybenzene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dimethoxybenzene
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.