missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, TRC

Product Code. 30387055
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387055 25 g -
30376456 100 g -
30376191 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387055 Supplier Toronto Research Chemicals Supplier No. TRCD45638025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-78-7
Molecular Formula C8 H10 O2
Molecular Weight (g/mol) 138.164
Synonym Benzene, p-dimethoxy- (8CI), 1,4-Dimethoxybenzene, 1,4-Dimethoxybenzol, 4-Methoxyanisole, DMB, Hydroquinone dimethyl ether, Methyl 4-methoxyphenyl ether, Methyl p-methoxyphenyl ether, NSC 7483, Quinol dimethyl ether, p-Dimethoxybenzene, p-Hydroquinone dimethyl ether, p-Methoxyanisole
IUPAC Name 1,4-dimethoxybenzene
SMILES COc1ccc(OC)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H10O2/c1-9-7-3-5-8(10-2)6-4-7/h3-6H,1-2H3
Formula Weight 138.0681
Chemical Name or Material 1,4-Dimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.