missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorobenzyl)-1,4-diazepane, 97%, Thermo Scientific™

Product Code. 10582452
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10582452 1 g 1g
10764657 5 g 5g
10525675 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10582452 Supplier Thermo Scientific Maybridge Supplier No. AW00512DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 76141-89-4
Molecular Formula C12H17FN2
Molecular Weight (g/mol) 208.28
MDL Number MFCD00816588
InChI Key KZCHYFUMQQWGJK-UHFFFAOYSA-N
Synonym 1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
PubChem CID 2774433
IUPAC Name 1-[(4-fluorophenyl)methyl]-1,4-diazepane
SMILES C1CNCCN(C1)CC2=CC=C(C=C2)F

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 208.28
Percent Purity 95%
Chemical Name or Material 1-(4-Fluorobenzyl)-1, 4-diazepane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.