missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%

Product Code. 11419487
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419487 5 g 5g
11429487 25 g 25g
11439487 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419487 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B22003.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52431-78-4 | C7H6N4OS | 194.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52431-78-4
Molecular Formula C7H6N4OS
Molecular Weight (g/mol) 194.212
MDL Number MFCD00132898
InChI Key MOXZSKYLLSPATM-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-5-mercaptotetrazole, 1-4-hydroxyphenyl-1h-tetrazole-5-thiol, 1-4-hydroxyphenyl-2h-tetrazole-5-thione, 1-4-hydroxyphenyl-5-mercapto-1h-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-4-hydroxyphenyl, 1-4-hydroxyphenyl-1h-tetrazole-5 4h-thione, 4-5-sulfanyl-1h-1,2,3,4-tetrazol-1-yl phenol, 1,2-dihydro-1-p-hydroxyphenyl-5h-tetrazole-5-thione, 4-5-mercapto-1h-tetrazol-1-yl phenol, 4-5-sulfanyl-1h-tetrazol-1-yl phenol
PubChem CID 3034725
IUPAC Name 1-(4-hydroxyphenyl)-2H-tetrazole-5-thione
SMILES C1=CC(=CC=C1N2C(=S)N=NN2)O

Specifications

Melting Point 169°C to 171°C
Quantity 5 g
Formula Weight 194.21
Percent Purity 96%
Chemical Name or Material 1-(4-Hydroxyphenyl)-5-mercaptotetrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.