missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%

Product Code. 11439487
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439487 100 g 100g
11419487 5 g 5g
11429487 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439487 Supplier Thermo Scientific Alfa Aesar Supplier No. B22003.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. Se returpolicy

CAS: 52431-78-4 | C7H6N4OS | 194.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 52431-78-4
Molecular Formula C7H6N4OS
Molecular Weight (g/mol) 194.212
MDL Number MFCD00132898
InChI Key MOXZSKYLLSPATM-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-5-mercaptotetrazole, 1-4-hydroxyphenyl-1h-tetrazole-5-thiol, 1-4-hydroxyphenyl-2h-tetrazole-5-thione, 1-4-hydroxyphenyl-5-mercapto-1h-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-4-hydroxyphenyl, 1-4-hydroxyphenyl-1h-tetrazole-5 4h-thione, 4-5-sulfanyl-1h-1,2,3,4-tetrazol-1-yl phenol, 1,2-dihydro-1-p-hydroxyphenyl-5h-tetrazole-5-thione, 4-5-mercapto-1h-tetrazol-1-yl phenol, 4-5-sulfanyl-1h-tetrazol-1-yl phenol
PubChem CID 3034725
IUPAC Name 1-(4-hydroxyphenyl)-2H-tetrazole-5-thione
SMILES C1=CC(=CC=C1N2C(=S)N=NN2)O

Specifikationer

Melting Point 169°C to 171°C
Quantity 100 g
Formula Weight 194.21
Percent Purity 96%
Chemical Name or Material 1-(4-Hydroxyphenyl)-5-mercaptotetrazole

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.