missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Hydroxyphenyl)piperazine, 98%, Thermo Scientific™

Product Code. 10582992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10582992 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10582992 Supplier Thermo Scientific Acros Supplier No. 367150010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56621-48-8
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C10H14N2O
MDL Number MFCD00066156
Quantity 1g
Synonym 1-4-hydroxyphenyl piperazine, 4-piperazin-1-yl phenol, 4-1-piperazinyl phenol, n-4-hydroxyphenyl piperazine, p-1-piperazinyl phenol, 4-piperazinophenol, phenol, 4-1-piperazinyl, 1-4-hydroxyphenyl-piperazine, 4-piperazinylphenol, 4-piperazinyl phenol
InChI Key GPEOAEVZTOQXLG-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)N1CCNCC1
IUPAC Name 4-(piperazin-1-yl)phenol
Molecular Weight (g/mol) 178.24
PubChem CID 92467
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material 1-(4-Hydroxyphenyl)piperazine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.