missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile, 96%

Product Code. 10414453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10414453 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10414453 Supplier Thermo Scientific Acros Supplier No. 170620250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16728-00-0 | C11H11NO | 173.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16728-00-0
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C11H11NO
Refractive Index 1.5415 to 1.5435
MDL Number MFCD00019212
Quantity 25 g
Specific Gravity 1.08
Synonym 1-4-methoxyphenyl cyclopropanecarbonitrile, 1-4-methoxyphenyl cyclopropane-1-carbonitrile, cyclopropanecarbonitrile, 1-4-methoxyphenyl, 1-4-methoxyphenyl-1-cyclopropanecarbonitrile, 1-4-methoxy-phenyl cyclopropanecarbonitrile, 4-1-cyanocyclopropyl anisole, 1-4-methoxy-phenyl-cyclopropanecarbonitrile, cyclopropanecarbonitrile,1-4-methoxyphenyl, 1-4-methoxyphenyl-cyclopropane-1-carbonitrile, 1-4-methoxyphenyl cyclopropane carbonitrile
InChI Key OIQRGAQXQLASRC-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C1(CC1)C#N
IUPAC Name 1-(4-methoxyphenyl)cyclopropane-1-carbonitrile
Molecular Weight (g/mol) 173.22
PubChem CID 85574
Formula Weight 173.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.