missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid, 96%

Product Code. 10429460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
Product Code. Quantity Packaging unitSize
10429460 10 g Glass bottle 10g
11955121 50 g Glass bottle 50g
2 options
This item is not returnable. View return policy

Product Code. 10429460

Brand: Thermo Scientific Acros 170710100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16728-01-1 | C11H12O3 | 192.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16728-01-1
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00019223
InChI Key WCPFQQHADRJANG-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl cyclopropanecarboxylic acid, 1-4-methoxyphenyl cyclopropane-1-carboxylic acid, 1-4-methoxyphenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-methoxyphenyl, 1-4-methoxy-phenyl-cyclopropanecarboxylic acid, 4-1-carboxycycloprop-1-yl anisole, 1-carboxy-1-4-methoxyphenyl cyclopropane, 1-4-methoxyphenyl cyclopropanecarboxylicacid, cyclopropanecarboxylicacid, 1-4-methoxyphenyl, acmc-1bprp
PubChem CID 85575
IUPAC Name 1-(4-methoxyphenyl)cyclopropane-1-carboxylic acid
SMILES COC1=CC=C(C=C1)C1(CC1)C(O)=O

Specifications

Melting Point 127.0°C to 132.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 192.21
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.