missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)piperazine, 98%

Product Code. 10712073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712073 5 g Glass bottle 5g
10722073 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712073 Supplier Thermo Scientific Acros Supplier No. 370830050
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39593-08-3 | C11H16N2 | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39593-08-3
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00040737
InChI Key ONEYFZXGNFNRJH-UHFFFAOYSA-N
Synonym 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
PubChem CID 83113
IUPAC Name 1-(4-methylphenyl)piperazine
SMILES CC1=CC=C(C=C1)N2CCNCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29°C to 34°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 176.26
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 1-(4-Methylphenyl)piperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(4-Methylphenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.