missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)phenyl]ethanol, 96%

Product Code. 11313816
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313816 5 g 5g
11303816 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313816 Supplier Thermo Scientific Alfa Aesar Supplier No. L16878.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1737-26-4 | C9H9F3O | 190.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1737-26-4
Molecular Formula C9H9F3O
Molecular Weight (g/mol) 190.165
MDL Number MFCD00039618
InChI Key YMXIDIAEXNLCFT-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl phenyl ethanol, 1-4-trifluoromethyl phenyl ethan-1-ol, a-methyl-4-trifluoromethylbenzyl alcohol, alpha-methyl-4-trifluoromethylbenzyl alcohol, alpha-methyl-4-trifluoromethyl benzyl alcohol, acmc-1boxb, 1-4-trifluoromethylphenyl-ethanol, 1-4-trifluoromethyl-phenyl-ethanol, alpha-methyl-4 trifluoromethyl benzyl alcohol
PubChem CID 137186
IUPAC Name 1-[4-(trifluoromethyl)phenyl]ethanol
SMILES CC(C1=CC=C(C=C1)C(F)(F)F)O

Specifications

Density 1.237
Boiling Point 62°C to 63°C (0.5 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.458
Quantity 5 g
Beilstein 2257808
Formula Weight 190.17
Percent Purity 97%
Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.