missing translation for 'onlineSavingsMsg'
Learn More

1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10793833
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10793833 1 g 1g
10213622 10 g 10g
11316965 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793833 Supplier Thermo Scientific Maybridge Supplier No. TL00212DA
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17610-21-8
Molecular Formula C16H22O
Molecular Weight (g/mol) 230.351
InChI Key IHUSZOMIBSDQTB-UHFFFAOYSA-N
Synonym 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one, 1,1,4,4-tetramethyl-6-acetyltetralin, 1-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethanone, 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-naphthalen-2-yl-ethanone, 6-acetyl-1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 2-acetyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 6-acetyl-1,1,4,4-tetramethyltetralin, 6-acetyl-1,1,4,4-tetra-methyl-tetralin, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydro-6-acetylnaphthalene
PubChem CID 2747562
IUPAC Name 1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C(CCC2(C)C)(C)C

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 230.35
Percent Purity 95%
Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.