missing translation for 'onlineSavingsMsg'
Learn More

1,5,5-Trimethylhydantoin, 98%

Product Code. 11421227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421227 5 g 5g
11431227 25 g 25g
11441227 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421227 Supplier Thermo Scientific Alfa Aesar Supplier No. B24188.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6851-81-6 | C6H10N2O2 | 142.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 6851-81-6
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.158
MDL Number MFCD00040439
InChI Key ZNYIPTYJBRGSSL-UHFFFAOYSA-N
Synonym 1,5,5-trimethylhydantoin, 3,4,4-trimethyl-2,5-dioxoimidazolidine, 1,5,5-trimethyl-imidazolidine-2,4-dione, 1,5,5-trimethyl-1,3-diazolidine-2,4-dione, hydantoin, 1,5,5-trimethyl, 2,4-imidazolidinedione, 1,5,5-trimethyl, 1,5,5 trimethylhydantoin, acmc-209o3u, 1,5,5-trimethyl-hydantoin, 1,5,5-trimethyl hydantoin
PubChem CID 81295
IUPAC Name 1,5,5-trimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)NC(=O)N1C)C

Especificaciones

Melting Point 161°C to 164°C
Quantity 5 g
Formula Weight 142.16
Percent Purity 98%
Chemical Name or Material 1,5,5-Trimethylhydantoin

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.