missing translation for 'onlineSavingsMsg'
Learn More

1,5-Anhydro-l-glucitol Bromhexine, TRC

Product Code. 30330595
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330595 50 mg 50mg
30348620 25 mg 25mg
30347659 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30330595 Supplier Toronto Research Chemicals Supplier No. TRCB67865750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,5-Anhydro-l-glucitol Bromhexine
Analyte or Component Names 1,5-Anhydro-l-glucitol Bromhexine
Shipping Condition Room Temperature
Molecular Formula C20 H30 Br2 N2 O5
Synonym (3S,4R,5R,6S)-2-((2,4-Dibromo-6-((cyclohexyl(methyl)amino)methyl)phenyl)amino)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
InChI Formula InChI=1S/C20H30Br2N2O5/c1-24(13-5-3-2-4-6-13)9-11-7-12(21)8-14(22)16(11)23-20-19(28)18(27)17(26)15(10-25)29-20/h7-8,13,15,17-20,23,25-28H,2-6,9-10H2,1H3/t15-,17-,18+,19-,20?/m0/s1
Solution Type Neat
SMILES CN(Cc1cc(Br)cc(Br)c1NC2O[C@@H](CO)[C@H](O)[C@@H](O)[C@@H]2O)C3CCCCC3
IUPAC Name (3S,4R,5R,6S)-2-[2,4-dibromo-6-[[cyclohexyl(methyl)amino]methyl]anilino]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 538.27
Quantity 50 mg
Formula Weight 536.0521
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.