Learn More
1,5-Diphenylcarbazide, 97+%
Beskrivelse
1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.
Solubility
Soluble in glacial acetic acid, alcohol and acetone. Slightly soluble in water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
Tekniske data
Tekniske data
| Melting Point | 170°C to 176°C |
| Quantity | 25 g |
| Beilstein | 752039 |
| Sensitivity | Air and light sensitive |
| Merck Index | 14,3321 |
| Solubility Information | Soluble in glacial acetic acid,alcohol and acetone. Slightly soluble in water. |
| Formula Weight | 242.28 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 1,5-Diphenylcarbazide |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.