missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbazide, 97+%

Product Code. 11497976
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497976 100 g 100g
11487976 25 g 25g
11407986 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497976 Supplier Thermo Scientific Alfa Aesar Supplier No. A14060.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 140-22-7 | C13H14N4O | 242.282 g/mol

1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

Solubility
Soluble in glacial acetic acid, alcohol and acetone. Slightly soluble in water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00003013
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 170°C to 176°C
Quantity 100 g
Beilstein 752039
Sensitivity Air and light sensitive
Merck Index 14,3321
Solubility Information Soluble in glacial acetic acid,alcohol and acetone. Slightly soluble in water.
Formula Weight 242.28
Percent Purity ≥97%
Chemical Name or Material 1,5-Diphenylcarbazide
compliance-icons
Product Identifier
  • 1,5-Diphenylcarbazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1,5-Diphenylcarbazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.