missing translation for 'onlineSavingsMsg'
Learn More

1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10106792
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10106792 1 g 1g
10350383 250 mg 250mg
10317164 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10106792 Supplier Thermo Scientific Maybridge Supplier No. KM07694DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 406476-31-1
Molecular Formula C12H13F3N2O2
Molecular Weight (g/mol) 274.24
MDL Number MFCD00209248
InChI Key KNDSIDUPVUCATQ-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylic acid, 1-5-trifluoromethyl-2-pyridyl piperidine-4-carboxylic acid, 5'-trifluoromethyl-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4-carboxylic acid, peakdale1_002677, 2-4-carboxypiperidin-1-yl-5-trifluoromethyl pyridine, 4-carboxy-1-5-trifluoromethyl pyridin-2-yl piperidine, 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylicacid, 1-5-trifluoromethylpyrid-2-yl-piperidine-4-carboxylic acid
PubChem CID 2777788
SMILES [O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid
Formula Weight 274.24
Percent Purity 95%
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridyl]piperidine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.