missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diphenoxy-2,4-hexadiyne, 98+%, Thermo Scientific™

Product Code. 11332397
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332397 25 g 25g
11322397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332397 Supplier Thermo Scientific Alfa Aesar Supplier No. L10004.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30980-37-1
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.308
MDL Number MFCD00041712
InChI Key ZBTRFBNXAKJTBI-UHFFFAOYSA-N
Synonym 1,6-diphenoxy-2,4-hexadiyne, 1,6-diphenoxyhexa-2,4-diyne, 6-phenoxyhexa-2,4-diyn-1-yl oxy benzene, 6-phenoxyhexa-2,4-diynyloxy benzene, maybridge1_002715, acmc-20ap13, 1,6-diphenoxy-2-4-hexadiyne
PubChem CID 1810326
IUPAC Name 6-phenoxyhexa-2,4-diynoxybenzene
SMILES C1=CC=C(C=C1)OCC#CC#CCOC2=CC=CC=C2

Specifications

Melting Point 80°C to 83°C
Linear Formula C6H5OCH2C≡C-C≡CCH2OC6H5
Quantity 25 g
Beilstein 1886340
Formula Weight 262.31
Percent Purity ≥98%
Chemical Name or Material 1,6-Diphenoxy-2,4-hexadiyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.