missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diphenyl-1,3,5-hexatriene, 95%

Product Code. 10704041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10704041 1 g Glass bottle 1g
10496720 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10704041 Supplier Thermo Scientific Acros Supplier No. 117300010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1720-32-7 | C18H16 | 232.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1720-32-7
Molecular Formula C18H16
Molecular Weight (g/mol) 232.32
MDL Number MFCD00004793
InChI Key BOBLSBAZCVBABY-WPWUJOAOSA-N
Synonym 1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
PubChem CID 5376733
ChEBI CHEBI:51594
IUPAC Name [(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
SMILES C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 199°C to 203°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCH=CHCH=CHC6H5
Quantity 1 g
Beilstein 05, 691
Formula Weight 232.32
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1,6-Diphenyl-1,3,5-hexatriene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.