missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediamine, 99.5+%

Product Code. 11353942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11353942 5 kg Plastic drum 5kg
10560551 100 g Plastic bottle 100g
10604782 500 g Plastic bottle 500g
10677652 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11353942 Supplier Thermo Scientific Acros Supplier No. 120640051
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-09-4 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine
SMILES C(CCCN)CCN

Specifications

Melting Point 38.0°C to 41.0°C
Color White
Boiling Point 204.0°C to 205.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Plastic drum
Linear Formula H2N(CH2)6NH2
Quantity 5 kg
Beilstein 04,269
Merck Index 15,4727
Solubility Information Solubility in water: 490g/L (20°C). Other solubilities: soluble in ethanol, benzene, diethyl ether
Formula Weight 116.21
Percent Purity 99.5+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1, 6-Hexanediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.