missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diaza-2,9-diketocyclotetradecane, TRC

Product Code. 30007496
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007496 2.5 mg 2.50mg
30012440 25 mg 25mg
30006864 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007496 Supplier Toronto Research Chemicals Supplier No. TRCD4176432.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56403-09-9
Molecular Formula C12H22N2O2
Molecular Weight (g/mol) 226.32
Synonym 1,8-Diaza-2,9-dioxocyclotetradecane, 1,8-Diazacyclopentadecane-2,9-dione, Hexahydro-2H-azepin-2-one Cyclic Dimer, 6-Aminohexanoate Cyclic Dimer, Caprolactam Cyclic Dimer;, Caprolactam Dimer, Cyclic Dimer of ε-Caprolactam, 6-Aminohexanoic Acid Bimol. Cyclic Lactam, ε-Aminocaproic Acid Cyclic Dimer, ε-Caprolactam Cyclic Dimer;
SMILES [H][C@@]1([C@@H](C(CC/C=C(C)\C)=C)CC2)[C@]2(C)[C@]3(C)CC[C@@]4([H])C(C)(C)[C@@H](O[C@]5([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@]6([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O6)CC[C@]4(C)[C@@]3([H])C[C@H]1O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C12H22N2O2/c15-11-7-3-1-5-9-13-12(16)8-4-2-6-10-14-11/h1-10H2,(H,13,16)(H,14,15)
Formula Weight 226.1681
Percent Purity >95%
Chemical Name or Material 1,8-Diaza-2,9-diketocyclotetradecane
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.