missing translation for 'onlineSavingsMsg'
Learn More

1.8-Diazacyclotetradecane-2,7-dione, TRC

Product Code. 30006626
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006626 10 mg 10mg
30005557 50 mg 50mg
30008849 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006626 Supplier Toronto Research Chemicals Supplier No. TRCD49480010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4266-66-4
Molecular Formula C12H22N2O2
Molecular Weight (g/mol) 226.32
Synonym NSC 138477
SMILES O=C1CCCCC(=O)NCCCCCCN1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H22N2O2/c15-11-7-3-4-8-12(16)14-10-6-2-1-5-9-13-11/h1-10H2,(H,13,15)(H,14,16)
Formula Weight 226.1681
Percent Purity >95%
Chemical Name or Material 1.8-Diazacyclotetradecane-2,7-dione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.