missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane

Product Code. 15447605
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447605 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447605 Supplier Thermo Scientific Alfa Aesar Supplier No. H26643.MD
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 214078-93-0 | C24H36N4 | 380.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 214078-93-0
Melting Point 67°C to 68°C
Molecular Formula C24H36N4
MDL Number MFCD09263312
Quantity 250 mg
Sensitivity Hygroscopic
Synonym 1,8-dibenzyl-1,4,8,11-tetraazacyclotetradecane, acmc-1cidi, n 1 ,n 8-dibenzylcyclam, 1,4,8,11-tetraazacyclotetradecane,1,8-bis phenylmethyl
InChI Key QWGFFCRTIFBAFJ-UHFFFAOYSA-N
SMILES C1CNCCN(CCCNCCN(C1)CC2=CC=CC=C2)CC3=CC=CC=C3
IUPAC Name 1,8-dibenzyl-1,4,8,11-tetrazacyclotetradecane
Molecular Weight (g/mol) 380.58
PubChem CID 11176466
Formula Weight 380.58
Chemical Name or Material 1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.