missing translation for 'onlineSavingsMsg'
Learn More

1'-Acetonaphthone, 95%

Product Code. 10386831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10386831 250 g 250g
10215041 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386831 Supplier Thermo Scientific Acros Supplier No. 400092500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 941-98-0 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-98-0
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00004013
InChI Key QQLIGMASAVJVON-UHFFFAOYSA-N
Synonym 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone
PubChem CID 13663
IUPAC Name 1-naphthalen-1-ylethanone
SMILES CC(=O)C1=CC=CC2=CC=CC=C21

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 12.0°C
Color Yellow
Density 1.1190g/mL
Boiling Point 302.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C10H7COCH3
Refractive Index 1.6270 to 1.6290
Quantity 250 g
Specific Gravity 1.119
Solubility Information Solubility in water: immiscible
Formula Weight 170.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1'-Acetonaphthone, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 1'-Acetonaphthone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.