missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxy-2-methoxybenzene, 98%

Product Code. 11499706
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499706 25 g 25g
11409716 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499706 Supplier Thermo Scientific Alfa Aesar Supplier No. B20242.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15212-03-0 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15212-03-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00017221
InChI Key BHJHPYFAYGAPLS-UHFFFAOYSA-N
Synonym guaiacol acetate, 1-acetoxy-2-methoxybenzene, guaiacyl acetate, o-acetoxyanisole, o-methoxyphenyl acetate, o-anisyl acetate, acetyl guaiacol, phenol, 2-methoxy-, acetate, o-acetylguaiacol, eucol
PubChem CID 61155
ChEBI CHEBI:86645
IUPAC Name (2-methoxyphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1OC

Specifications

Quantity 25 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 1-Acetoxy-2-methoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.