missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-2-naphthol, 99%

Product Code. 11408457
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408457 25 g 25g
11329609 5 g 5g
11418457 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408457 Supplier Thermo Scientific Alfa Aesar Supplier No. A18537.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 574-19-6 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 574-19-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00012131
InChI Key VUIOUIWZVKVFCI-UHFFFAOYSA-N
Synonym 2'-hydroxy-1'-acetonaphthone, 1-acetyl-2-naphthol, 1-2-hydroxynaphthalen-1-yl ethanone, 1-2-hydroxy-1-naphthalenyl ethanone, 1-2-hydroxy-1-naphthyl ethan-1-one, 2-hydroxy-1-acetonaphthone, 1-2-hydroxy-1-naphthyl ethanone, ethanone, 1-2-hydroxy-1-naphthalenyl, 1-acetyl-2-hydroxynaphthalene, 1'-acetonaphthone, 2'-hydroxy
PubChem CID 68455
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)ethanone
SMILES CC(=O)C1=C(C=CC2=CC=CC=C21)O

Specifications

Melting Point 63°C to 67°C
Boiling Point 130°C to 132°C (2 mmHg)
Quantity 25 g
Beilstein 1869900
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 1-Acetyl-2-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.