missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylnaphthalene, 97+%

Product Code. 11489733
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489733 50 g 50g
11499733 250 g 250g
11479733 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489733 Supplier Thermo Scientific Alfa Aesar Supplier No. A12918.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 941-98-0 | C12H10O | 170.211 g/mol

They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-98-0
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004013
InChI Key QQLIGMASAVJVON-UHFFFAOYSA-N
Synonym 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone
PubChem CID 13663
IUPAC Name 1-naphthalen-1-ylethanone
SMILES CC(=O)C1=CC=CC2=CC=CC=C21

Specifications

Melting Point 9°C to 11°C
Density 1.12
Boiling Point 302°C
Flash Point >110°C (230°F)
Refractive Index 1.628
Quantity 50 g
Beilstein 1100618
Solubility Information Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.
Formula Weight 170.21
Percent Purity ≥97%
Chemical Name or Material 1-Acetylnaphthalene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Acetylnaphthalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-Acetonaphthone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.