missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-propanol, Dr. Ehrenstorfer

Product Code. 30319273
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319273 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319273 Supplier Dr. Ehrenstorfer Supplier No. DRECA10216950
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Amino-2-propanol
CAS 78-96-6
Analyte or Component Names 1-Amino-2-propanol
Shipping Condition +20°C
Molecular Formula C3 H9 N O
Certifications/Compliance ISO/IEC 17025
Synonym 1-Amino-2-propanol, (2-Hydroxy-2-methylethyl)amine, (RS)-1-Amino-2-hydroxypropane, (RS)-1-amino-2-propanol, (±)-1-Amino-2-propanol, 1-Amino-2-hydroxypropane, 1-Aminoisopropanol, 1-Methyl-2-aminoethanol, 2-Amino-1-methylethanol, 2-Hydroxy-1-propanamine, 2-Hydroxy-1-propylamine, 2-Hydroxypropanamine, 2-Hydroxypropylamine, DL-1-Amino-2-propanol, Isopropanolamine, MIPA, Monoisopropanolamine, NSC 3188, Threamine, dl-1-Amino-2-propanol, α-Aminoisopropyl alcohol, β-Aminoisopropanol
InChI Formula InChI=1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3
SMILES CC(O)CN
IUPAC Name 1-aminopropan-2-ol
Molecular Weight (g/mol) 75.11
Quantity 500 mg
Formula Weight 75.0684 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.