missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyrene, 97%

Product Code. 10341134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341134 1 g Glass bottle 1g
10408820 250 mg Glass bottle 250mg
15109153 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10341134 Supplier Thermo Scientific Acros Supplier No. 104530010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1606-67-3 | C16H11N | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1606-67-3
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.27
MDL Number MFCD00004140
InChI Key YZVWKHVRBDQPMQ-UHFFFAOYSA-N
Synonym 1-aminopyrene, 1-pyrenamine, 3-aminopyrene, pyrene, amino, pyren-1-ylamine, aminopyrene, pyrenamine, alpha-aminopyrene, unii-luw9eo1681, ccris 760
PubChem CID 15352
IUPAC Name pyren-1-amine
SMILES NC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Beilstein 12,1341
Formula Weight 217.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Aminopyrene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Aminopyrene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.