missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydryl-3-azetidinone, 95%, Thermo Scientific™

Product Code. 15474358
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474358 500 mg 500mg
15464358 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474358 Supplier Thermo Scientific Alfa Aesar Supplier No. H57966.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40320-60-3
Molecular Formula C16H15NO
Molecular Weight (g/mol) 237.302
MDL Number MFCD00964539
InChI Key AVUDXLOVIBJFQA-UHFFFAOYSA-N
Synonym 1-diphenylmethyl azetidin-3-one, 1-benzhydryl-azetidin-3-one, 1-1,1-diphenylmethyl azetidin-3-one, 1-diphenylmethyl-3-azetidinone, 1-benzhydryl-3-azetidinone, 3-azetidinone, 1-diphenylmethyl, pubchem10151, acmc-209jd2, n-benzhydrylazetidin-3-one
PubChem CID 3735558
IUPAC Name 1-benzhydrylazetidin-3-one
SMILES C1C(=O)CN1C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Density 1.182
Boiling Point 350°C to 355°C
Flash Point 156°C (313°F)
Refractive Index 1.626
Quantity 500 mg
Formula Weight 237.3
Percent Purity 95%
Chemical Name or Material 1-Benzhydryl-3-azetidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.