missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylazetan-3-ol, Thermo Scientific™

Product Code. 10602073
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602073 1 g Amber Glass Bottle 1g
10069321 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10602073 Supplier Thermo Scientific Maybridge Supplier No. KM08723DA
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 18621-17-5
Molecular Formula C16H17NO
Molecular Weight (g/mol) 239.32
MDL Number MFCD00205109
InChI Key MMAJXKGUZYDTHV-UHFFFAOYSA-N
Synonym 1-benzhydrylazetan-3-ol, 1-diphenylmethyl-3-hydroxyazetidine, 1-diphenylmethyl azetidin-3-ol, 1-benzhydryl-3-azetidinol, 1-benzhydryl-3-hydroxyazetidine, 1-benzhydryl-azetidin-3-ol, n-benzhydrylazetidin-3-ol, 3-azetidinol, 1-diphenylmethyl, 1-diphenylmethyl-3-azetidinol
PubChem CID 330448
SMILES OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 1-benzhydrylazetidin-3-ol
Formula Weight 239.32
Percent Purity 97%
Chemical Name or Material 1-Benzhydrylazetan-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.