missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylazetane-3-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10612643
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10612643 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10612643 Supplier Thermo Scientific Maybridge Supplier No. KM10090DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 36476-87-6
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C17H17NO2
MDL Number MFCD01566539
Quantity 1 g
Synonym 1-diphenylmethyl azetidine-3-carboxylic acid, 1-benzhydrylazetane-3-carboxylic acid, 1-benzhydryl-3-azetidinecarboxylic acid, 3-azetidinecarboxylic acid, 1-diphenylmethyl, 1-benzhydryl-azetidine-3-carboxylic acid, 1-diphenylmethylazetidine-3-carboxylic acid, 1-benzhydryl-3-azetanecarboxylic acid, 1-diphenylmethyl-3-azetidinecarboxylic acid, maybridge3_004072, 1-benzhydrylazetidine-3-carboxylicacid
InChI Key BRSCYENHLCPOAU-UHFFFAOYSA-N
SMILES C1C(CN1C(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
IUPAC Name 1-benzhydrylazetidine-3-carboxylic acid
Molecular Weight (g/mol) 267.328
PubChem CID 2735440
Formula Weight 267.33
Percent Purity 95%
Chemical Name or Material 1-Benzhydrylazetane-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.