missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylpiperazine, 97%

Product Code. 11382467
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11382467 25 g 25g
11392467 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11382467

Brand: Thermo Scientific Alfa Aesar L11530.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 841-77-0 | C17H20N2 | 252.361 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 841-77-0
Molecular Formula C17H20N2
Molecular Weight (g/mol) 252.361
MDL Number MFCD00038379
InChI Key NWVNXDKZIQLBNM-UHFFFAOYSA-N
Synonym 1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
PubChem CID 70048
IUPAC Name 1-benzhydrylpiperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 91°C to 92°C
Quantity 25 g
Beilstein 222773
Sensitivity Air Sensitive
Formula Weight 252.36
Percent Purity 97%
Chemical Name or Material 1-Benzhydrylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.