missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-L-tryptophan, 97%

Product Code. 15436048
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436048 5 g 5g
15446048 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436048 Supplier Thermo Scientific Alfa Aesar Supplier No. H59663.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143824-78-6 | C31H30N2O6 | 526.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143824-78-6
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153366
InChI Key ADOHASQZJSJZBT-SANMLTNESA-N
Synonym fmoc-trp boc-oh, fmoc-l-trp boc-oh, nalpha-9h-fluoren-9-ylmethoxy carbonyl-n1-tert-butoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1-tert-butoxycarbonyl-1h-indol-3-yl propanoic acid, 2s-3-1-tert-butoxycarbonyl indol-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-trp boc, pubchem10048, 1-boc-n-fmoc-l-tryptophan, ksc910o8t, n1-boc-nalpha-fmoc-l-tryptophan
PubChem CID 9849766
SMILES CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Melting Point ∽97°C (decomposition)
Quantity 5 g
UN Number UN3077
Beilstein 7698009
Sensitivity Moisture sensitive
Optical Rotation −21° (c=1 in DMF)
IUPAC Name 3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Formula Weight 526.58
Percent Purity 97%
Chemical Name or Material 1-Boc-N-Fmoc-L-tryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.