missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-piperazine, 99%

Product Code. 11388076
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388076 5 g 5g
11378076 1 g 1g
11398076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388076 Supplier Thermo Scientific Alfa Aesar Supplier No. L13363.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 57260-71-6 | C9H19N2O2 | 187.26 g/mol

1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.

Solubility
Soluble in ethyl acetate, methanol and water.

Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57260-71-6
Molecular Formula C9H19N2O2
Molecular Weight (g/mol) 187.26
MDL Number MFCD00075265
InChI Key CWXPZXBSDSIRCS-UHFFFAOYSA-O
Synonym 1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
PubChem CID 143452
IUPAC Name tert-butyl piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC[NH2+]CC1

Specifications

Melting Point 44°C to 48°C
Quantity 5 g
Beilstein 879985
Sensitivity Air Sensitive
Solubility Information Soluble in ethyl acetate,methanol and water.
Formula Weight 186.26
Percent Purity 99%
Chemical Name or Material 1-Boc-piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.