missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dichlorobenzene, 98%

Product Code. 11455033
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455033 5 g 5g
11465033 25 g 25g
11475033 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455033 Supplier Thermo Scientific Alfa Aesar Supplier No. A12298.06
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1193-72-2 | C6H3BrCl2 | 225.894 g/mol

1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-72-2
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00018333
InChI Key ISHYFWKKWKXXPL-UHFFFAOYSA-N
Synonym 2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l
PubChem CID 70947
IUPAC Name 1-bromo-2,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Br

Specifications

Melting Point 26°C to 27°C
Boiling Point 235°C
Quantity 5 g
Beilstein 1635733
Solubility Information Slightly soluble in water.
Formula Weight 225.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,4-dichlorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.