missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dimethoxybenzene, 98%, Thermo Scientific™

Product Code. 10522921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10522921 5 mL Glass bottle 5mL
10255280 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10522921 Supplier Thermo Scientific Acros Supplier No. 134170050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17715-69-4
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00009844
InChI Key NIUZVSQOXJIHBL-UHFFFAOYSA-N
Synonym 2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
PubChem CID 87266
IUPAC Name 1-bromo-2,4-dimethoxybenzene
SMILES COC1=CC=C(Br)C(OC)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 25°C to 26°C
Color Yellow
Density 1.5070g/mL
Boiling Point 153°C to 155°C (18.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OCH3)Br
Refractive Index 1.5718 to 1.5738
Quantity 5 mL
Beilstein 06, II, 819
Specific Gravity 1.507
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 217.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 4-dimethoxybenzene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.