missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-difluoro-4-nitrobenzene, 97%, Thermo Scientific Chemicals

Product Code. 11462958
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462958 5 g 5g
11452958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462958 Supplier Thermo Scientific Alfa Aesar Supplier No. A18670.06
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167415-27-2
Molecular Formula C6H2BrF2NO2
Molecular Weight (g/mol) 237.99
MDL Number MFCD00042184
InChI Key GJFYMYJYPARISZ-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoronitrobenzene, 4-bromo-2,5-difluoro-1-nitrobenzene, benzene, 1-bromo-2,5-difluoro-4-nitro, buttpark 29\01-58, 1-bromo-2,5-difluoro-4-nitro-benzene, pubchem8498, zlchem 1046, acmc-209dwg, intermediates-zcf02256, ksc495e4h
PubChem CID 2736286
IUPAC Name 1-bromo-2,5-difluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(Br)C=C1F

Specifications

Melting Point 55°C to 57°C
Quantity 5 g
Beilstein 7411347
Formula Weight 237.99
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,5-difluoro-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.