missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-difluorobenzene, 98%, Thermo Scientific™

Product Code. 10442811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10442811 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10442811 Supplier Thermo Scientific Acros Supplier No. 249370050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 399-94-0
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H3BrF2
Linear Formula FC6H3BrF
Refractive Index 1.5075 to 1.5095 (20°C, 589nm)
MDL Number MFCD00000345
Quantity 5g
Synonym 1-bromo-2,5-difluorobenzene, 2,5-difluorobromobenzene, benzene, 2-bromo-1,4-difluoro, 1,4-difluoro-2-bromobenzene, 2-bromo-1,4-difluoro-benzene, fr df be, pubchem2302, acmc-1ad5i, 1-bromo-2,5 difluorobenzene, 2,5-difluoro-1-bromobenzene
InChI Key XCRCSPKQEDMVBO-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)Br)F
IUPAC Name 2-bromo-1,4-difluorobenzene
Molecular Weight (g/mol) 192.991
PubChem CID 67862
Formula Weight 192.99
Percent Purity ≥97.5%
Chemical Name or Material 1-Bromo-2, 5-difluorobenzene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.