missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-iodobenzene, 98+%, ACROS Organics™

Product Code. 10698254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10698254 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10698254 Supplier Thermo Scientific Acros Supplier No. 295430050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 136434-77-0
CAS Min % 98.0
CAS Max % 100.0
Melting Point 36°C to 41°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C6H3BrFI
Linear Formula BrC6H3(F)I
MDL Number MFCD00042577
Quantity 5 g
Synonym 4-bromo-3-fluoroiodobenzene, 3-fluoro-4-bromo iodobenzene, 1-bromo-2-fluoro-4-iodo-benzene, benzene, 1-bromo-2-fluoro-4-iodo, 3-fluoro-4-bromo-iodobenzene, 1-bromo-2-fluoro-4-i, pubchem3231, acmc-209c5x, 3-fluoro-4-bromoiodobenzene, ksc490o1h
InChI Key OCODJNASCDFXSR-UHFFFAOYSA-N
SMILES FC1=C(Br)C=CC(I)=C1
IUPAC Name 1-bromo-2-fluoro-4-iodobenzene
Molecular Weight (g/mol) 300.90
PubChem CID 2725006
Formula Weight 300.89
Percent Purity ≥98.0%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1-Bromo-2-fluoro-4-iodobenzene
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.